missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ MultiTube Vortexers: Test Tube Racks

Test tube racks for Fisherbrand™ MultiTube Vortexers

Brand:  Fisherbrand™

Additional Details : Weight : 0.07000kg

 View more versions of this product

Product Code. 15569004

  • £32.20 / Each
Estimated Shipment: 30-08-2024
to see stock.
Add to basket

Description

Description

Test tube racks for Fisherbrand™ MultiTube Vortexers

For use with Fisher Scientific MultiTube Vortexers

Color-coded foam racks for test tubes

Specifications

Specifications

100 mL
Gray
50
Test Tube Rack
Fisher Scientific MultiTube Vortexer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers