Learn More
Fisherbrand™ Test Tube Holders for Heavy-duty Vortex Mixer
For use with heavy-duty vortex mixer.
Brand:  Fisherbrand™ 80863056
 View more versions of this product
Product Code. 15539034
  • £52.00 / Each
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Holds a wide range of test tube numbers and diameters
  • Requires Universal Holder
Specifications
Specifications
Test Tube Holder
Heavy-duty vortex mixer
4
26 to 29 mm
Foam insert
Universal holder