missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Traceable™ Digital Thermometer

Versatile panel-mount module snaps into place or can be used as a stand-alone unit.

Brand:  Fisherbrand™ 93677-20

49.03 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.


Product Code. 11789735

  • £56.93 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Includes:
  • Stainless-steel probe with extended cable
  • Individually serial-numbered
  • Battery

Description

Description

  • Easy-view, four-digit LCD is 0.75 in. high
  • Records minimum and maximum memory readings over any time period
  • Temperature range encompasses virtually all testing requirements: -58° to +572°F and -50 to +300°C Resolution is 0.1° from -20° to +200°; 1° outside this range
  • Accuracy is ±1°C between -20° and +100°C
  • Reading updates every second
  • Display switches between English and Metric units

Order Info

Dimensions: 1.5H x 2.44W x 0.5 in. D, Probe diameter 0.14 in. , Probe length (including handle): 2.75 in. , Cable length: 10

Certifications

Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.

Specifications
Show More
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Traceable™ Digital Thermometer > Digital thermometer; Snap-in Module with Probe; Range: -50° to +300°C (-58° to +572°F

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.