Learn More
Includes: Stainless steel probe; 9V Alkaline battery; High-impact, chemical-resistant ABS plastic case
Description
Provides routine measurements with semisolid, liquid, or air/gas. Fisherbrand™ Traceable™ Digital Thermometers can switch between Celsius and Fahrenheit temperatures. Easy-to-read LCD screen includes Min/Max memory. Switch-selectable temperature ranges: -50° to +150°C and -58° to +302°F.
- Universal, stainless-steel 316 probe is triple purpose for liquid, air/gas, and semisolids.
- Ideal for foods, pharmaceuticals, petroleum products, gas systems, soil, ovens, and more
- Unit reads in °C and °F and includes a HOLD key for freezing data on the display for future readings
- Instant HISTORY key recalls minimum and maximum readings over the past 24 hours
- Unit conforms to ITS-90 International Temperature Standard
Order Info
Order additional probes separately; contact your Customer Service Representative. Accessory adapter also available.
Certifications
Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™; indicates traceability to standards provided by NIST™.
Specifications
ISO 17025, A2LA, NIST | |
32 mm | |
Measuring temperatures in cuvettes, gas systems, reagents, chemical solutions, soil, water baths, ovens, incubators, pharmaceuticals, petroleum products, foods and wastewater | |
140 mm | |
59 in. | |
29 cm | |
152.4 mm | |
Stainless Steel with USB Connector | |
8 oz. | |
3.5 in. | |
9 in. | |
150°C | |
-40°C | |
-40°C to +150°C | |
±0.05°C | |
89 mm |
1.26 in. | |
0.5 in.H (13 mm) | |
5.5 in. | |
Stainless steel probe with USB connector; 9V Alkaline battery; High-impact, chemical-resistant ABS plastic case | |
6 in. | |
150 cm | |
ABS Plastic | |
0.0001°C | |
226.8 g | |
Digital Thermometer | |
300°F | |
-40°F | |
-40°F to +300°F | |
Yes | |
Digital LCD |
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85% -
Multiple Manufacturers
Your input is important to us. Please complete this form to provide feedback related to the content on this product.