Learn More
Fisherbrand™ Traceable™ Digital Thermometers with Short Sensors
Fisherbrand™ Traceable™ Digital Thermometers are ideal for monitoring liquids, air/gas or semisolids.
Brand:  Fisherbrand™ 4047
 View more versions of this product
Product Code. 11735843
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Choose from models with a variety of features including MAX/MIN function to record the maximum and minimum temperature in a certain time period; visual and audible alarms and multiple displays of recorded temperatures.
  • Select the correct temperature to use in water baths, refrigerators, incubators and tanks
Certifications
Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™ indicates traceability to standards provided by NIST™.
Specifications
Specifications
ISO 17025, A2LA, NIST
2.25 x 3.25 x 2.25 in. (5.7 x 8.2 x 5.71 cm)
2.25 in.
Sensor, AAA alkaline battery, flip open stand, wall mount, magnet and Velcro™ mounting tape
0.8 in.
305 cm
0.1°C
70.8 g
Digital Thermometer: Dual Scale
158°F
-58°F
-58°F to +158°F
±1°C
8.2 cm
0.18 in.
Monitoring liquids, air/gas or semisolids in water baths, refrigerators, freezers, incubators and tanks
5.71 cm
10 ft.
57 mm
2.03 cm
2.5 oz.
3.25 in.
2.25 in.
70°C
-50°C
-50°C to +70°C
Digital LCD