missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Traceable™ Platinum Ultra-Accurate Replacement Handle Probe is an accessory for use with Traceable Platinum Ultra-Accurate digital Thermometer.
- High-accuracy: 0.05 percent between -80 to +290 deg. Celsius
- Waterproof
- 4-wire platinum/stainless-steel
Specifications
0.13 in. | |
6 ft. | |
1.8 m (Cable), 17.8 cm (Probe) | |
17.8 cm | |
Platinum RTD stainless-steel Probe | |
932°F | |
392°F | |
-328°F to +932°F | |
Traceable Platinum Ultra-Accurate digital Thermometer |
3.3 mm | |
7 in. | |
1.8 m | |
Platinum, Stainless Steel | |
Replacement Handle Probe | |
500°C | |
200°C | |
-200°C to +500°C | |
0.05 percent between -80 to +290 deg.C |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Traceable™ Platinum Ultra-Accurate Replacement Handle Probe > Traceable™ Platinum Ultra-Accurate Replacement Handle Probe
For improved performance, probes should be calibrated as a system with the meter.
Spot an opportunity for improvement?Share a Content Correction