Learn More
Thermo Scientific™ Wellwash™ and Wellwash™ Versa™ Microplate Strip Washer: Bottles
Accessories for Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
Brand:  Thermo Scientific™ N10806
 View more versions of this product
Product Code. 11727996
  • £211.00 / Each
Explore more special offers
Description
Description
Accessories for Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
For use with Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
  • Subsitute bottles of different sizes for added convenience
  • Wash and waste bottles include liquid level sensors
  • 4L wash and 9L waste bottles available for automation applications
ELISA-based assays, cell washing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cap with tubing and liquid level sensors
2 L
Wellwash Microplate Strip Washer
67.62 oz.
Waste Bottle