Learn More
Thermo Scientific™ Wellwash™ and Wellwash™ Versa™ Microplate Strip Washer: Bottles
Accessories for Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
£211.00 - £552.00
Product Code | Brand | Capacity (Metric) | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Type | Price | Quantity & Availability | ||||
11708006
|
Thermo Scientific™
N10812 |
4 L | Wash Bottle B |
£367.00
Each |
Estimated Shipment: 09-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11737996
|
Thermo Scientific™
N10810 |
4 L | Waste Bottle |
£244.00
Each |
Estimated Shipment: 14-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11797996
|
Thermo Scientific™
N10811 |
4 L | Wash Bottle A |
£367.00
Each |
Estimated Shipment: 09-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11718006
|
Thermo Scientific™
N10813 |
4 L | Rinse Bottle |
£367.00
Each |
Estimated Shipment: 09-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11758006
|
Thermo Scientific™
N10814 |
9 L | Waste Bottle |
£552.00
Each |
Estimated Shipment: 14-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11777996
|
Thermo Scientific™
N10809 |
2 L | Rinse Bottle |
£257.00
Each |
Estimated Shipment: 09-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11767996
|
Thermo Scientific™
N10808 |
2 L | Wash Bottle B |
£258.00
Each |
Estimated Shipment: 14-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11727996
|
Thermo Scientific™
N10806 |
2 L | Waste Bottle |
£211.00
Each |
Estimated Shipment: 14-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11747996
|
Thermo Scientific™
N10805 |
2 L | Wash Bottle |
£253.00
Each |
Estimated Shipment: 14-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11757996
|
Thermo Scientific™
N10807 |
2 L | Wash Bottle A |
£257.00
Each |
Estimated Shipment: 14-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Accessories for Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
For use with Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
- Subsitute bottles of different sizes for added convenience
- Wash and waste bottles include liquid level sensors
- 4L wash and 9L waste bottles available for automation applications
ELISA-based assays, cell washing
Specifications
Cap with tubing and liquid level sensors |