Learn More
Thermo Scientific™ Wellwash™ and Wellwash™ Versa™ Microplate Strip Washer: Bottles
Accessories for Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
Brand: Thermo Scientific™ N10807
Descripción
Accessories for Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
For use with Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
- Subsitute bottles of different sizes for added convenience
- Wash and waste bottles include liquid level sensors
- 4L wash and 9L waste bottles available for automation applications
ELISA-based assays, cell washing
Especificaciones
Cap with tubing and liquid level sensors | |
2 L | |
Wellwash Versa Microplate Strip Washer |
67.62 oz. | |
Wash Bottle A |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.