Learn More
Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents
Minimize the risks of improper measurement and contamination with these chemistry reagents, which are contained in self-filling vials.
Brand: Thermo Scientific™ AC4099
Description
Minimize the risks of improper measurement and contamination with Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents, which are contained in self-filling vials. Use these reagents with the Orion™ AQUAfast AQ4000 colorimeter to simplify sample preparation and tests. The auto-test ampoules contain a pre-measured amount of liquid reagent and easily fill with the correct volume of sample for simple and fast sample and reagent mixing. Each ampoule contains an Auto-ID barcode that is read by the AQ4000 colorimeter and used to automatically identify the test parameter and initiates the test.
- No. of test: 30
- Consistency and reproducibility improved with AQUAfast ampoules and an AQ4000 meter
- Thermo Scientific™ Auto-ID™ eliminates measurement errors

Specifications
Chlorine Dioxide | |
Orion AQUAfast AQ4000 Colorimeter | |
30 Auto-ID Ampoules | |
Colorimetric water and wastewater testing equipment | |
0.8 to 11 mg/L ClO2 | |
30 Ampoules |
Ampoule | |
AQUAfast chlorine dioxide Auto-ID ampoules, 30 pack | |
DPD | |
AQUAfast Zero Vial Auto-Test Kit (Cat. No. AQ4ZER) | |
Drinking Water: Standard Methods 4500 ClO2 D |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- Chlorine Dioxide
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P370+P378-In case of fire: Use CO2, dry chemical, or foam to extinguish.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : acetone
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.