Learn More
Thermo Scientific™ Wellwash™ and Wellwash™ Versa™ Microplate Strip Washer: Bottles
Accessories for Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
Brand: Thermo Scientific™ N10813
Description
Accessories for Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
For use with Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
- Subsitute bottles of different sizes for added convenience
- Wash and waste bottles include liquid level sensors
- 4L wash and 9L waste bottles available for automation applications
ELISA-based assays, cell washing
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
135.25 oz. | |
Round | |
Wellwash Versa Microplate Strip Washer |
4 L | |
Rinse Bottle |