missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Wellwash™ and Wellwash™ Versa™ Microplate Strip Washer: Bottles
Accessories for Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
Brand: Thermo Scientific™ N10810
This item is not returnable.
View return policy
Description
Accessories for Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
For use with Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
- Subsitute bottles of different sizes for added convenience
- Wash and waste bottles include liquid level sensors
- 4L wash and 9L waste bottles available for automation applications
ELISA-based assays, cell washing

Specifications
Cap with tubing and liquid level sensors | |
4 L | |
Waste Bottle |
135.25 oz. | |
Square | |
Wellwash Microplate Strip Washer, Wellwash Versa Microplate Strip Washer |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Wellwash™ and Wellwash™ Versa™ Microplate Strip Washer: Bottles > 4L waste bottle, square
Spot an opportunity for improvement?Share a Content Correction