missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Propellers for ServoDyne™ Electronic Mixers
Three-bladed propeller for general purpose mixing
Brand: Cole-Parmer™ 04552-10
This item is not returnable.
View return policy
Description
- Can be used as single or mounted several on a shaft for thorough agitation
- Electro-polished
- Marine-style propellers: used for general-purpose mixing
- Turbine style propellers: used for airless mixing in deep containers
Specifications
3-blade Propeller | |
0.78 cm | |
5.1 cm | |
ServoDyne™ electronic mixer |
0.31 in. | |
2 in. | |
Ideal for airless mixing in deep containers |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cole-Parmer™ Propellers for ServoDyne™ Electronic Mixers > Turbine style; Overall diameter: 2 in. (51mm)
Spot an opportunity for improvement?Share a Content Correction