Learn More
Fisherbrand™ Combs for FB-VE16-1 Vertical Electrophoresis System
Creates the wells in gels for electrophoresis
Brand:  Fisherbrand™ P1-24C
 View more versions of this product
Product Code. 11903055
  • £41.15 / Each
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Owl P1-CST Multiple Gradient Gel Caster
16 x 14 cm
0.08 cm
7.87 in.
20 cm
5.5 in.
13 cm
0.29 cm
18 cm
Comb
24
51 μL
6.3 in.
16 cm
0.11 in.
14 cm
7.08 in.