missing translation for 'onlineSavingsMsg'
Learn More
Fisherbrand™ Easy-Squeeze Wash Bottles, 6/Cs. Product Code.: 11775233
Add to basket

Fisherbrand™ Easy-Squeeze Wash Bottles, 6/Cs.
GREENER_CHOICE

Product Code. 11775233 Shop All Fisherbrand Products
Blue, Distilled Water
Click to view available options
Color:
Assorted
Blue
Green
Natural
Red
Yellow
Chemical Label:
Acetone
Assorted Cleaning
Assorted Solvents
Deionized Water
Distilled Water
Ethanol
Isopropanol
Methanol
Saline
Soap
Sodium Hypochlorite (Bleach)
Universal
Water
Wash bottles in this range that are prelabelled for intended usage with specific hazardous substances are also prelabelled with the appropriate GHS hazard warning symbols.
 

Product Code. 11775233

Add to basket
Add to basket
Wash bottles in this range that are prelabelled for intended usage with specific hazardous substances are also prelabelled with the appropriate GHS hazard warning symbols.
 

Includes

Come with colored polypropylene closures with PTFE membrane (except 02-897-2, which has an HDPE closure).

Easy to fill and ready to use

  • Wide mouth
  • Translucent and sputter-free
  • With easy-to-read color-coded silk-screened labels
  • Tips can be cut back to increase flow
  • When not in use, bottles relieve internal pressure without dripping

Label information includes:

  • Chemical name
  • National Fire Protection Association (NFPA) codes and symbols
  • Chemical Abstracts Service (CAS) number
  • Hazard codes
  • Laboratory protective equipment information
  • Target organ information
  • Recommended fill level
  • LDPE recycling code
  • Bottle volume capacity “For Dispensing Only, Not Recommended For Storage” statement

Not recommended for use above 60°C (140°F).

Order Info

Available in cases of six, all with the same label; in a six-bottle Solvent Pack (one each labeled acetone, deionized water, ethanol, isopropanol, methanol and universal); or six-bottle Cleaning Pack (one each labeled acetone, bleach, deionized water, saline, soap and universal bottles).

Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
TRUSTED_SUSTAINABILITY

Specifications

Capacity 17 oz., 500mL
Capacity (Metric) 500 mL
Capacity (English) 17 oz.
Color Blue
Diameter (English) 2.99 in.
Includes Color-coded Silk screened labels, PTFE membrane
Material LDPE
Closure Color Blue
Certifications/Compliance ISO 13485:2003
Closure Included Yes
Autoclavable Not Autoclavable
Graduated No
Mouth Wide
Shape Cylindrical
Height (Metric) 173 mm
Product Type Wash Bottle
Height (English) 6.81 in.
Chemical Label Distilled Water
Closure Material Polypropylene
Closure Size 38-415 mm
Diameter (Metric) 76 mm
Closure Type Polypropylene Closure
Through the Closure Dispensing No
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Easy-Squeeze Wash Bottles, 6/Cs. >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.