missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ 96-Well Plates
For EIA and other applications
£241.28 - £241.49
Specifications
Format | Standard |
---|---|
Array | 8 x 12 |
Binding Type | High Binding |
Color | Clear |
Product Type | 96-well Assay Microplate |
Product Code | Brand | Certifications/Compliance | Sterility | Well Shape | Lid | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Certifications/Compliance | Sterility | Well Shape | Lid | Price | Quantity & Availability | ||
11997924
|
Fisherbrand™
|
Certified | Non-sterile | Rounded Square | Without Lid |
£241.49
Pack of 50 |
In Stock
Log in to see stock. |
||
11917924
|
Fisherbrand™
|
Not certified | Sterile | Rounded Square | With Lid |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
For EIA and other applications
- Clear polystyrene
- Results with CV of <5% between wells and replicate values within ±10% of the mean Background of wells is within 0.005 absorbance units from mean Small lip at each well top and ridges between wells prevent well-to-well contamination Flat well bottom surface provides greater area for binding
- Rows and columns are labeled alphanumerically; plates stack for easy storage
- Specially treated to ensure high binding of hydrophobic or partially hydrophobic molecules like viral antigens and proteins
Order Info
Nonsterile microplates are supplied in bulk packs of 50 plates. Sterile microplates come individually wrapped with lids in packs of 40 plates.
Certifications
Cat. No. 21-377-203 is supplied with a certificate attesting to its high-binding capabilities.
Specifications
Standard | |
High Binding | |
96-well Assay Microplate | |
450 μL | |
Rounded Square |
8 x 12 | |
Clear | |
Polystyrene | |
96 | |
EIA and other applications |