missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Absorption Macro Rectangular Cells with PTFE Lid

Fit standard spectrophotometer cell holders

Brand:  Fisherbrand™ 14958120

 View more versions of this product

Product Code. 15276023

  • £126.00 / Pack of 2
Estimated Shipment: 20-02-2025
to see stock.
Explore more special offers
Add to basket

Includes: PTFE lid

Description

Description

Fit standard spectrophotometer cell holders

  • All cells are 12.5mm wide
  • 1, 5 or 10mm path length rectangular cells
  • Available with a variety of window materials for wide range of assays
  • Available in four materials e.g. Quartz, Optical Quartz, Special Optical Glass and Optical Glass
  • Quartz windows for transmission from far ultraviolet through the infrared
  • Glass windows for routine work in ultraviolet region and visible region

Specifications

Specifications

Macro Absorption Cell
Optical Glass
10mm
10 mm
45 mm
12.5 mm
Rectangle
3.5 mL
PTFE
2/Pk., 8 Pk./Cs.
360 to 2500 nm
Standard Spectrophotometer Cell Holder
Routine work in ultraviolet region and visible region
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.