missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Absorption Quartz Micro Rectangular Cells with PTFE Lid

Fit standard spectrophotometer cell holders

Brand:  Fisherbrand™ 14958133

 View more versions of this product

Product Code. 15256073


Includes: PTFE lid
This item is not returnable. View return policy

Description

Description

Fit standard spectrophotometer cell holders

  • All cells are 12.5mm wide
  • 10mm path length rectangular cells
  • Reduce reagent use with Micro cells
  • Quartz windows for transmission from far ultraviolet through the infrared

Specifications

Specifications

Micro Absorption Cell
Quartz
2mm
2/Pk., 8 Pk./Cs.
200 to 2500 nm
Standard Spectrophotometer Cell Holder
Transmission from far ultraviolet through the infrared
300 μL
PTFE
2 clear windows
10 mm
25 mm
12.5 mm
Rectangle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers