missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Absorption Ultra Micro Rectangular Cells with PTFE Lid
Fit standard spectrophotometer cell holders
Brand: Fisherbrand™ 14958138
Includes: PTFE lid or stopper
Description
- All cells are 12.5mm wide
- 1, 5 or 10mm path length rectangular cells
- Reduce reagent use with Ultramicro cells
- Quartz windows for transmission from far ultraviolet through the infrared
- Glass windows for routine work in ultraviolet region and visible region
Specifications
Ultra-micro Absorption Cell | |
50 μL | |
PTFE | |
Ea., 8 Ea./Cs. | |
200 to 2500 nm | |
Standard spectrophotometer cell holder | |
Transmission from far ultraviolet through the infrared |
Center:8.5mm | |
Quartz | |
2mm | |
10 mm | |
45 mm | |
12.5 mm | |
Rectangle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction