missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Absorption Ultra Micro Rectangular Cells with PTFE Lid

Fit standard spectrophotometer cell holders

Brand:  Fisherbrand™ 14958136

 View more versions of this product

Product Code. 15246103

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers

Includes: PTFE lid or stopper
This item is not returnable. View return policy

Description

Description

  • All cells are 12.5mm wide
  • 1, 5 or 10mm path length rectangular cells
  • Reduce reagent use with Ultramicro cells
  • Quartz windows for transmission from far ultraviolet through the infrared
  • Glass windows for routine work in ultraviolet region and visible region

Specifications

Specifications

Ultra-micro Absorption Cell
100 μL
PTFE
Ea., 8 Ea./Cs.
200 to 2500 nm
Standard spectrophotometer cell holder
Transmission from far ultraviolet through the infrared
Center:8.5mm
Quartz
2mm
10 mm
45 mm
12.5 mm
Rectangle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.