Learn More
Bio Plas™ Astral™ Inoculation Device System
Economical, disposable inoculation system designed for safety and dependable sterility
£643.00
Specifications
Disposable | Single-use |
---|---|
For Use With (Equipment) | Astral™ inoculation device system |
Product Code | Brand | Color | Product Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Product Type | Price | Quantity & Availability | ||||
11534000
|
Bio Plas™
7000 |
Yellow | Inoculating Loop | N/A | |||||
Description
Economical, disposable inoculation system designed for safety and dependable sterility
Calibrated using Evans Blue Dye spectrophotometric procedure and gravimetric method
- Free of DNA, RNase, DNase, pyrogens and PCR inhibitors
- Astral loops, needles, and Clone-Piks are packaged static free into individual tubular chambers, which are then hermetically sealed with aluminum film
- A dust cover is heat-shrunk onto the unit before it is irradiation sterilized
- Use on an individual basis, or for quantitative procedures such as sampling, urine counts, or serial dilutions
- Puncture aluminum film with the handle, pushing down until it attaches to the loop, needle, or Clone-Pik
- Withdraw handle; inoculation device is ready for many applications
Order Info
Supplied 10 racks of 96 to a case
Specifications
Single-use | |
Astral™ inoculation device system |