missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Autoclavable Biohazard Waste Bags
Dual tested compliant.
£86.16 - £422.00
Product Code | Brand | Dimensions (L x W) | Capacity (English) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Capacity (English) | Price | Quantity & Availability | ||||
18558056
|
Fisherbrand™
|
23 x 19 in. (58.4 x 48.3 cm) | 7 to 10 gal. |
£86.16
Pack of 100 |
Estimated Shipment: 06-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
18306513
|
Fisherbrand™
|
35 x 25 in. (89 x 63.5 cm) | 13 to 16 gal. |
£154.26
Pack of 100 |
Estimated Shipment: 17-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
18336533
|
Fisherbrand™
|
38 x 31 in. (96.5 x 78.7 cm) | 20 to 30 gal. |
£422.00
Pack of 200 |
Estimated Shipment: 17-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
18366523
|
Fisherbrand™
|
30 x 24 in. (76.2 x 61 cm) | 8 to 12 gal. |
£194.18
Pack of 200 |
Estimated Shipment: 17-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
18316533
|
Fisherbrand™
|
48 x 37 in. (122 x 94 cm) | 44 gal. |
£296.09
Pack of 100 |
Estimated Shipment: 17-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Dual tested compliant
Autoclaving is a very common way to sterilize infectious waste through the use of high temperature, high pressure steam over a specific time period. Sterilized waste is less expensive to dispose. Dual-tested autoclave bags are ASTM D1922 Elmendorf Tear tested to exceed 480g of tear strength (MD and TD) and ASTM D1709 tested to exceed 165g dart drop strength.- Integral steam processing indicator that turns brown when autoclaved
- Bags comply with US DOT 49CFR Sections 173-197 and specific state regulations
- Operating temperature tolerance up to 285degreeFdegree
- Available in various sizes
- Bag with black print
- Gauge: 2mil
Specifications
Autoclavable | |
Red with Black Print |