Learn More
Fisherbrand™ Cooling Block
Fisherbrand Aluminum alloy cooling block keeps samples in place prior and during freezing process.
Brand:  Fisherbrand™ 13131012
 View more versions of this product
Product Code. 16100381
  • £177.00 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Specifications
1 to 2 mL cryogenic vials
Aluminum
null
Cryo Block
-122.8°F
119 x 101 x 38 mm
12.2 cm
Black
Rectangular
30 (1 to 2 mL Cryogenic Vials)
1
-86°C
16.3 cm
4.3 cm