missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ PurSwab Foam Swabs

Flexible foam applicators. Tip is comprised of an open-cell polyurethane foam. Ideal for a variety of cleaning applications.

Brand:  Fisherbrand™ 149603G

30.35 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.


 View more versions of this product

Product Code. 16458115

  • £39.40 / Pack of 50
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Flexible mini-tip foam applicators. Tip is comprised of an open-cell polyurethane foam. Ideal for a variety of cleaning applications.

Cleaning sensors and components.  Soft non-abrasive foam tip.  Wood handle naturally biodegradable.  Made in USA
Specifications

Specifications

ISO 9001:2008, ISO 13485:2003, FDA GMP's
6 in.
White
0.211cm
5.812 in.
0.68 in.
0.478 cm
Medical
Wood
Polyurethane Foam
0.083 in.
Cleaning sensors and components, Applying adhesives or other solutions, Removing dirt, contamination, fluxes or resins, Soft non-abrasive foam tip, Wood handle is naturally biodegradable
Swab
15.2cm
14.76 cm
1.74 cm
0.188 in.
Non-sterile
Standard
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.