Learn More
Fisherbrand™ Magnetic Stirring Bars
Stirbar assortments come in a convenient storage box. Fisherbrand™ Magnetic Stirring Bars contain PTFE encapsulated Alnico magnets and come in various sizes.
Brand:  Fisherbrand™ 04775-99
 View more versions of this product
Product Code. 11716408
  • £300.00 / Pack of 12
Estimated Shipment: 12-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Stirbar assortments come in a convenient storage box. Fisherbrand™ Magnetic Stirring Bars contain PTFE encapsulated Alnico magnets and come in various sizes.
  • Choose from Octagon, Cylindrical, Polygon, Micro, Egg-Shaped or Spinplus™ shaped assortments
  • FDA Grade, USP Class VI PTFE low friction coating; durable and inert
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclavable
Specifications
Specifications
Autoclavable
White
PTFE
Octagon
ISO 9001:2008
Two each of the following sizes 0.5 x 0.31 in., 0.63 x 0.31 in., 1 x 0.38 in., 1.5 x 0.38 in., 2 x 0.38 in., and 2.5 x 0.38 in.
Alnico Magnet
Stir Bar