missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Autoclavable HD Biohazard Waste Bags

Biohazard symbol printed bags to ensure safe handling and disposal of biohazardous waste.

Brand:  Fisherbrand™ 01815A

 View more versions of this product

Product Code. 18386533

  • £40.40 / Pack of 100

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Impact- and tear-resistant autoclave bags for waste sterilization. Fisherbrand™ Orange Autoclave Bags without Sterilization Indicator are constructed of high-density polyethylene.

  • Impact- and tear-resistant.
  • Bright orange color.
  • High-density polyethylene construction

Compliance

Pass ASTM™ impact- and tear-resistance tests.

Specifications

Specifications

ASTM
Orange with Black Print
61 cm
Autoclavable
12 in.
38 micron
127°C
None
24 x 12 in. (61 x 30 cm)
30 cm
Printed in English
24 in.
HDPE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.