missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Autoclavable HD Biohazard Waste Bags
Biohazard symbol printed bags to ensure safe handling and disposal of biohazardous waste.
£40.40 - £93.90
Specifications
Certifications/Compliance | ASTM |
---|---|
Closure Type | None |
Color | Orange with Black Print |
Autoclavable | Autoclavable |
Includes | Printed in English |
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
18386533
|
Fisherbrand™
01815A |
24 x 12 in. (61 x 30 cm) |
£40.40
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18356523
|
Fisherbrand™
01815C |
35 x 24 in. (88.9 x 61 cm) |
£93.90
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10440633
|
Fisherbrand™
01815B |
30 x 24 in. (76 x 61 cm) |
£51.00
Pack of 100 |
Estimated Shipment: 10-04-2025
Log in to see stock. |
|||||
Description
Impact- and tear-resistant autoclave bags for waste sterilization. Fisherbrand™ Orange Autoclave Bags without Sterilization Indicator are constructed of high-density polyethylene.
- Impact- and tear-resistant.
- Bright orange color.
- High-density polyethylene construction
Compliance
Pass ASTM™ impact- and tear-resistance tests.
Specifications
ASTM | |
Orange with Black Print | |
Printed in English | |
HDPE |
None | |
Autoclavable | |
38 micron | |
127°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title