Learn More
Fisherbrand™ Autoclavable HD Biohazard Waste Bags
Biohazard symbol printed bags to ensure safe handling and disposal of biohazardous waste.
Brand: Fisherbrand™ 01815B
Description
Impact- and tear-resistant autoclave bags for waste sterilization. Fisherbrand™ Orange Autoclave Bags without Sterilization Indicator are constructed of high-density polyethylene.
- Impact- and tear-resistant.
- Bright orange color.
- High-density polyethylene construction
Compliance
Pass ASTM™ impact- and tear-resistance tests.
Specifications
ASTM | |
Orange with Black Print | |
76 cm | |
Autoclavable | |
24 in. | |
38 micron | |
127°C |
None | |
30 x 24 in. (76 x 61 cm) | |
61 cm | |
Printed in English | |
30 in. | |
HDPE |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.