missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Autoclavable HD Biohazard Waste Bags
Strong and leak-resistant
Brand: Fisherbrand™
Additional Details : Weight : 0.01100kg
Description
Strong and leak-resistant
- Printed with biohazard symbol and text in English and Spanish
- Supplied with twist tie closure
Specifications
Twist Tie | |
36 x 45 in. (91 x 114 cm) | |
91 cm | |
Biohazard symbol and text in English and Spanish | |
36 in. | |
Polyethylene |
Red with Black Print | |
114 cm | |
Autoclavable | |
45 in. | |
1.50 mil |