missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Polypropylene Graduated Cylinders
30.30 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Description
Calibrated To Contain/To Deliver at 20°C
- Translucent cylinders
- Molded-in large, easy-to-read graduations
- Large rounded base ribbed for reinforcement and stability
- Generous pour spout; 10mL size has a conical top for easier filling and pouring
- Non-wetting
- Chemical resistant
- Contact-clear
Specifications
2000 mL | |
Autoclavable | |
Circular |
20 mL | |
20mL | |
Graduated Cylinder |