Learn More
Hellma™ Quartz, Silica and Glass Standard Cell Cuvettes
Designed to hold samples for spectroscopic experiments and optical analysis. Hellma™ Quartz, Silica and Glass Standard Cell Cuvettes provide optically excellent transparency.
Brand:  Hellma™ 100-10-46
 View more versions of this product
Product Code. 10340721
  • £218.00 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Includes: Fitted PTFE covers or tapered PTFE stoppers, except 14-385-906A and -902A, which come with glass cover plates
Description
Description
  • Tried and tested over decades, used in countless applications
  • Utmost accuracy in terms of optical path length and parallelism
  • Very high temperature and chemical resistance due to thermal bonding of individual components
  • Unique surface flatness of the optical windows of 0.001mm
  • Function-optimized design with beveled edges and corners protects against the risk of damage caused by splitting and assists users in their daily work
Specifications
Specifications
Macro Cell 100-QX
3500 μL
9.5mm
10 mm
45 mm
2 windows
Quartz Suprasil
PTFE cover
200 to 3500 nm
12.5 mm