missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hellma™ Quartz, Silica and Glass Standard Cell Cuvettes
Designed to hold samples for spectroscopic experiments and optical analysis. Hellma™ Quartz, Silica and Glass Standard Cell Cuvettes provide optically excellent transparency.
£99.30 - £332.00
Includes: Fitted PTFE covers or tapered PTFE stoppers, except 14-385-906A and -902A, which come with glass cover plates
Product Code | Brand | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Price | Quantity & Availability | ||||||
10340721
|
Hellma™
100-10-46 |
£204.00
Each |
In Stock
Log in to see stock. |
||||||
11791754
|
Hellma™
100-5-40 |
With Suprasil quartz windows; Lightpath: 5mm; PTFE cover |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11771754
|
Hellma™
100-1-40 |
With Suprasil quartz windows; Lightpath: 1mm; Glass cover |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11781784
|
Hellma™
110-10-40 |
With Suprasil quartz windows; Lightpath: 10mm; PTFE stopper |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12711675
|
Hellma™
6030-UV-10-53E |
With standard silica windows; 2/pk. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Quartz - high UV transmission and low thermal expansion
- Glass - used in the visible range of the spectrum
- Low values of refractive index and dispertion
- Good resistance to chemicals