Learn More
Fisherbrand™ Sav-It™ Tube Closures
GREENER_CHOICE
Firm, leak-resistant seals protect personnel and samples
Brand:  Fisherbrand™ A21068FIS
 View more versions of this product
Product Code. 15364719
  • £33.85 / Pack of 1000
Explore more special offers
Description
Description
  • One-hand application
  • Can be punctured for through-cap sampling via closed-tube instrumentation systems
  • Won't interfere with most common chemistry, coagulation, and drug monitoring methodologies
  • Fit test tubes, centrifuge tubes, specialty tubes, and evacuated blood drawing tubes
  • Direct through-cap sampling via closed-tube instrumentation systems
  • Won't crack or fall off
  • Hold fast under most rigorous procedures including agitation, centrifugation, refrigeration, and freezing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Not Autoclavable
Green
12 to 13 mm
Test tubes, centrifuge tubes, specialty tubes, evacuated blood drawing tubes
Elastomer Copolymer
0.47 to 0.51 in.
Non-sterile