Learn More
Fisherbrand™ Screw Caps for Disposable Glass Tubes
Description
Phenolic caps have cemented-in rubber liners
- Autoclavable
- Polypropylene cap is linerless
Specifications
13-415 | |
No | |
Autoclavable | |
Black | |
14-959-35C |
Rubber | |
Screw Cap | |
Phenolic | |
Non-sterile |
Product Content Correction
Product Title
Fisherbrand™ Screw Caps for Disposable Glass Tubes > Phenolic; 13mm-415
Spot an opportunity for improvement?Share a Content Correction