Learn More
Fisherbrand™ Screw Caps for Disposable Glass Tubes
Phenolic caps have cemented-in rubber liners
Brand:  Fisherbrand™ FS73800-18415
 View more versions of this product
Product Code. 11567133
  • £174.00 / Pack of 1000
Estimated Shipment: 19-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Phenolic caps have cemented-in rubber liners
  • Autoclavable
  • Polypropylene cap is linerless
TRUSTED_SUSTAINABILITY
Specifications
Specifications
18-415
No
Autoclavable
Black
14-959-37A,14-957-76E, 14-959-37C
Rubber
Screw Cap
Phenolic
Non-sterile