missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Sterile Microcentrifuge Tubes with Screw Caps
Note: Image is representative only. It shows colored caps, but all of the tubes in this offering come with Natural caps; there are no color cap options.
Brand: Fisherbrand™ 02681374
440.04 GBP valid until 2024-12-31
Use promo code "21938" to get your promotional price.
This item is not returnable.
View return policy
Description
Choose skirted or conical-bottom style
- With attached caps
- Natural-colored screw caps with O-rings
- RNase-free and DNase-free
Specifications
18,000 x g | |
Autoclavable | |
Natural | |
O-ring | |
Screw Cap |
Polypropylene | |
2 mL | |
Polypropylene | |
Sterile | |
Skirted |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction