Learn More
Fisherbrand™ SureOne™ Reload Empty Racks
For use with Fisherbrand SureOne universal pipet tips
Brand: Fisherbrand™
12.65 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : A0
Additional Details : Weight : 0.76140kg
Description
- Inserts (not included) sit flush with rack, allowing easy multichannel pipetting
- Notched clasp designed to prop lid open for autoclave venting
- Sure stack lid rim prevents racks from sliding around when stacked and carted around a lab
- Safe lock latch allows for one-handed opening and closing ability—won't pop open when dropped
- Sure snap design securely holds insert in place, preventing it from being lifted out while pipetting
- Flip-lid can be either a hinged or lift-off lid; hinged format does not prevent use of a multichannel pipet
Order Info
Inserts not included; must be ordered separately.
Specifications
Empty Rack | |
Standard Pipette Tips (1250μL) |