missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Traceable™ Hi-Accuracy Thermometer

Monitors inlet and outlet simultaneously. Traceable™ Hi-Accuracy Thermometer records time and date for MIN/MAX readings.

Brand:  Fisherbrand™ 93676-51

 View more versions of this product

Product Code. 12950831


Includes: Probe(s), two AA batteries, Stand, Velcro™ magnetic strips, wall mount, Traceable™ certificate
This item is not returnable. View return policy

Description

Description

  • Two visual (LEDs) and two audio alarm alerts
  • Alarms programmable in 0.1° increments
  • Probe tip penetration of 0.33 in.
  • Stainless-steel probe with 0.125 in. diameter, 6.25 in. stem length, 9 in. overall length and 5 ft. cable length
  • Available with one probe or two probes

Certifications

Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.

Compliance

Conforms to ITS-90, International Temperature Standard

Specifications

Specifications

NIST, ITS-90, ISO 17025, A2LA
1.9 cm
3.05 mm
Measuring temperatures in water baths, cuvets, gas systems, reagents, incubators, pharmaceuticals, petroleum products, foods, wastewater
10.79 cm
5 ft.
230 mm
15.87 cm
5 oz.
2.75 in.
9 in.
70°C
-50°C
-50°C to +70°C
±0.3°C
6.98 cm
0.75 in.
0.12 in.
9 x 2.75 x 4.25 in. (22.86 x 6.98 x 10.79 cm)
4.25 in.
Stainless Steel Probe, Two AA Batteries, Stand, Velcro™ Magnetic Strips, Wall Mount, Traceable™ Certificate
6.25 in.
152.4 cm
0.01°C
141.7 g
Traceable Thermometer
158°F
-58°F
-58°F to +158°F
AA
LED
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers