missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Traceable™ Hi-Accuracy Thermometer
Monitors inlet and outlet simultaneously. Traceable™ Hi-Accuracy Thermometer records time and date for MIN/MAX readings.
£135.00 - £187.00
Specifications
Certifications/Compliance | NIST, ITS-90, ISO 17025, A2LA |
---|---|
Depth (English) | 0.75 in. |
Depth (Metric) | 1.9 cm |
Diameter (English) Probe | 0.12 in. |
Diameter (Metric) Probe | 3.05 mm |
Includes: Probe(s), two AA batteries, Stand, Velcro™ magnetic strips, wall mount, Traceable™ certificate
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
12950831
|
Fisherbrand™
93676-51 |
Stainless Steel Probe, Two AA Batteries, Stand, Velcro™ Magnetic Strips, Wall Mount, Traceable™ Certificate | |||||||
11761445
|
Fisherbrand™
93676-56 |
Two Stainless Steel Probes, Two AA Batteries, Stand, Velcro™ Magnetic Strips, Wall Mount, Traceable™ Certificate |
£187.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Two visual (LEDs) and two audio alarm alerts
- Alarms programmable in 0.1° increments
- Probe tip penetration of 0.33 in.
- Stainless-steel probe with 0.125 in. diameter, 6.25 in. stem length, 9 in. overall length and 5 ft. cable length
- Available with one probe or two probes
Certifications
Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.
Compliance
Conforms to ITS-90, International Temperature Standard
Specifications
NIST, ITS-90, ISO 17025, A2LA | |
1.9 cm | |
3.05 mm | |
Measuring temperatures in water baths, cuvets, gas systems, reagents, incubators, pharmaceuticals, petroleum products, foods, wastewater | |
10.79 cm | |
6.25 in. | |
152.4 cm | |
0.01°C | |
141.7 g | |
Traceable Thermometer | |
158°F | |
-58°F | |
-50°C to +70°C | |
±0.3°C | |
6.98 cm |
0.75 in. | |
0.12 in. | |
9 x 2.75 x 4.25 in. (22.86 x 6.98 x 10.79 cm) | |
4.25 in. | |
5 ft. | |
230 mm | |
15.87 cm | |
5 oz. | |
2.75 in. | |
9 in. | |
70°C | |
-50°C | |
AA | |
LED |