Learn More
Fisherbrand™ Traceable™ Vaccine Refrigerator/Freezer Thermometer
Monitors temperature of vaccine in freezers, water baths, heating blocks, incubators, and refrigerators with enclosed temperature-buffered sensor.
Brand: Fisherbrand™ 90008-35
93.10 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
Monitors temperature of vaccine in freezers, water baths, heating blocks, incubators, and refrigerators with enclosed temperature-buffered sensor. Fisherbrand™ Traceable™ Vaccine Refrigerator/Freezer Thermometer includes a probe which is sealed in glycol solution and insulates sensor when refrigerator door is opened.
- Triple display simultaneously shows Min/max and current temperatures, °F/°C switchable
- Bottle insulates sensor from rapid temperature changes when refrigerator door is opened
- 15-077-720 has a 5mL bottle designed for vaccines; 06-664-11 has a 10mL bottle
- Glass bottle filled with glycol mimics vaccine temperature and fits in trays
- Probe is sealed in glycol solution which insulates sensor when refrigerator door is opened
- Ten-foot micro-cable permits refrigerator doors to close on it
- Minimum/maximum memory monitors high and low readings overnight, on weekends, or for any time period
- Alarm is programmable in 1° increments, sounds continuously when temperature rises above/falls below set points and can be turned off manually
- High-impact, chemical-resistant ABS plastic case
- Temperature range: -50 to +70°C/-58 to +158°F
- Resolution: 1° accuracy: ±1C
- Battery:AA Alkaline (1)
Certifications
Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA. It provides traceability to standards provided by NIST.
Specifications
Glycol | |
ISO 17025, A2LA, NIST | |
Monitoring temperature in freezers, water baths, heating blocks, incubators and refrigerators | |
19 mm | |
10 in. | |
25.4 cm | |
Refrigerator/Freezer Thermometer with 10mL bottle | |
4 oz. | |
2.75 in. | |
2.75 in. | |
70°C | |
-50°C | |
-50°C to +70°C | |
Refrigerators/Freezers | |
±1°C | |
70 mm |
10 mL | |
2.75 x 2.75 x 0.75 in. (69.85 x 70 x 19 mm) | |
0.75 in. | |
Velcro™ and magnetic mounting strip, one AA battery, ABS Plastic case, alarm set in 1° increments alerts user with visual and audible signals if temperature goes out | |
70 mm | |
ABS Plastic | |
1°C | |
0.11 kg | |
Fridge/Freezer Digital Thermometer | |
158°F | |
-58°F | |
-58°F to +158°F | |
Yes | |
AA Alkaline (1) | |
Digital LCD |
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Your input is important to us. Please complete this form to provide feedback related to the content on this product.