Learn More
Novus Biologicals™ FITC-geldanamycin
Brand: Novus Biologicals™ NB610-81561
Additional Details : Weight : 0.00970kg
Specifications
Store at -20 C. Avoid freeze-thaw cycles. | |
Purple Solid. Soluble in DMSO (>10 mg/mL). | |
Geldanamycin |
1 mg | |
FITC-geldanamycin | |
FITC |