missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ FiveEasy™ F30 Conductivity Meters
Compact, benchtop meter provides high quality conductivity measurements with simple click of a button
£785.00 - £935.00
Specifications
Accuracy (Conductivity) | ±0.5% of the measured value |
---|---|
Calibration | One point calibration with three predefined standards |
Range (Conductivity) | 0.01 μS/cm to 200 mS/cm |
Resolution (Conductivity) | Automatic range |
Temperature Compensation | Linear: 0.00%/°C to 10.00%/°C, reference temperature: 20° and 25°C |
Product Code | Brand | Type | Includes | For Use With (Application) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Includes | For Use With (Application) | Price | Quantity & Availability | |||
15503380
|
Mettler Toledo™
30266942 |
F30 Meter Only | F30 pH meter, electrode holder, 1 each buffers 1413mS/cm and 12.88mS/cm standard sachets, test certificate | Applications that require a specific sensor not included in a kit |
£785.00
1 unit |
Estimated Shipment: 11-09-2024
Log in to see stock. |
|||
15513380
|
Mettler Toledo™
30266943 |
F30 Standard Kit | F30 pH meter, LE703 pH sensor, electrode holder, 1 each buffers 1413mS/cm and 12.88mS/cm standard sachets, test certificate | Suited for aqueous samples with mid to high conductivity |
£935.00
1 unit |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||
Description
- Compact, space saving design with ABS housing
- Remove sensor holder after use and store on the side of the meter
- Large, well-structured 4.3 in. segmented LCD display for easy viewing
- Intuitive button arrangement and simple menu for easy operation
- One point calibration with three predefined standards
- Mini-DIN sensor input
- Features Automatic and manual endpoint recording, Acoustic endpoint signal, Visual endpoint signal
- Ideal for wide range of laboratory applications
- Conductivity Range: 0.01μS/cm to 200mS/cm
- Conductivity Resolution: Automatic range
- Conductivity Accuracy: ±0.5% of the measured value
- TDS Range: 0.01mg/L to 200g/L
- TDS Resolution: Automatic range
- TDS Accuracy: ±0.5% of the measured value
- Temperature Range: 0° to 100°C (32° to 212°F)
- Temperature Resolution: 0.1°C
- Temperature Accuracy: 0.5°C
- Temperature Compensation: Linear: 0.00%/°C to 10.00%/°C, reference temperature: 20° and 25°C
- Memory Size: current calibration
- Power Supply: 100 to 240V, 50/60Hz, 12 VDC
- Dimensions: 22.7 x 14.7 x 7cm (9 x 5.7 x 2.7 in.)
- Weight: 0.63kg (1.4 lb.)
- F30 Meter only: Suited for applications that require a specific sensor not included in a kit
- F30 Standard Kit: Suited for aqueous samples with mid to high conductivity
Specifications
±0.5% of the measured value | |
0.01 μS/cm to 200 mS/cm | |
Linear: 0.00%/°C to 10.00%/°C, reference temperature: 20° and 25°C | |
0.1°C | |
Current calibration | |
22.7 x 14.7 x 7 cm (9 x 5.7 x 2.7 in.) | |
1.4 lb. | |
50/60Hz |
One point calibration with three predefined standards | |
Automatic range | |
32°F to 212°F | |
0.5°C | |
Mini-DIN | |
0.63 kg | |
100/240V |