missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ FiveEasy™ Plus FP20 pH/mV Meters

Compact, benchtop meter provides high quality pH/mV measurements with simple click of a button.

Brand:  Mettler Toledo™ 30266627

571.47 GBP valid until 2024-12-31
Use promo code "21937" to get your promotional price.


 View more versions of this product

Product Code. 15553360

  • £715.00 / 1 unit
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket

Includes: Meter and charger only. (Does not include electrode holder, buffers or probe.)
This item is not returnable. View return policy

Description

Description

  • Compact, space saving design with ABS housing
  • Large, well-structured 4.3 in. segmented LCD display for easy viewing
  • Intuitive button arrangement and simple menu for easy operation
  • RS232 or USB ports to export data directly either to a printer or a PC for further processing
  • ATC and MTC temperature compensation
  • Five point (max.) linear only calibration with four predefined buffer groups
  • BNC, Cinch, NTC 30kiloOhm sensor inputs
  • Features Automatic and manual endpoint recording, Acoustic endpoint signal, Visual endpoint signal
  • Ideal for wide range of laboratory applications
Specifications:
  • pH Range: -2.00 to 16.00
  • pH Resolution: 0.01/0.1
  • pH Accuracy: ±:0.01
  • mV Range: 2000 to 2000mv
  • mV Resolution: 1mV
  • mV Accuracy: ±1mV
  • Temperature Range: -5° to +105°C (23° to 221°F)
  • Temperature Resolution: 0.1°C
  • Temperature Accuracy: 0.3°C
  • Memory Size: 200 measurements, current calibration
  • Power Supply: 100 to 240V, 50/60Hz, 12 VDC
  • Dimensions: 22.7 x 14.7 x 7cm (9 x 5.7 x 2.7 in.)
  • Weight: 0.63kg (1.4 lb.)
Applications:
  • FP20 Meter only: Suited for applications that require a specific sensor not included in a kit
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FiveEasy Plus FP20-Basic
0.3°C
±0.01
Applications that require a specific sensor not included in a kit
-2.00 to 16.00
2000 to 2000
-5°C to +105°C
0.1°C
200 measurements, current calibration
22.7 x 14.7 x 7 cm (9 x 5.7 x 2.7 in.)
1.4 lb.
FP20 Meter Only
50/60 Hz
FP20 pH meter, charger, test certificate, pH measurement guide
±1
RS232, USB
Linear/Segmented
0.01, 0.1
1mV
23°F to 221°F
LCD
BNC, Cinch, NTC 30kiloOhm sensors
0.63 kg
ATC and MTC
120/240 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.