missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ FiveEasy™ Plus FP20 pH/mV Meters

Compact, benchtop meter provides high quality pH/mV measurements with simple click of a button.

Brand:  Mettler Toledo™ 30266941

581.25 GBP valid until 2024-12-15
Use promo code "21658" to get your promotional price.


 View more versions of this product

Product Code. 15593370

  • £886.00 / 1 unit
Estimated Shipment: 06-12-2024
to see stock.
Explore more special offers
Add to basket

Includes: LE420 sensor, installation QuickGuide, pH buffer starter pack
This item is not returnable. View return policy

Description

Description

  • Compact, space saving design with ABS housing
  • Remove sensor holder after use and store on the side of the meter
  • Large, well-structured 4.3 in. segmented LCD display for easy viewing
  • Intuitive button arrangement and simple menu for easy operation
  • RS232 or USB ports to export data directly either to a printer or a PC for further processing
  • ATC and MTC temperature compensation
  • Five point (max.) linear only calibration with four predefined buffer groups
  • BNC, Cinch, NTC 30kiloOhm sensor inputs
  • Features Automatic and manual endpoint recording, Acoustic endpoint signal, Visual endpoint signal
  • Ideal for wide range of laboratory applications
Specifications:
  • pH Range: -2.00 to 16.00
  • pH Resolution: 0.01/0.1
  • pH Accuracy: ±:0.01
  • mV Range: 2000 to 2000mv
  • mV Resolution: 1mV
  • mV Accuracy: ±1mV
  • Temperature Range: -5° to +105°C (23° to 221°F)
  • Temperature Resolution: 0.1°C
  • Temperature Accuracy: 0.3°C
  • Memory Size: 200 measurements, current calibration
  • Power Supply: 100 to 240V, 50/60Hz, 12 VDC
  • Dimensions: 22.7 x 14.7 x 7cm (9 x 5.7 x 2.7 in.)
  • Weight: 0.63kg (1.4 lb.)
Applications:
  • FP20 Meter only: Suited for applications that require a specific sensor not included in a kit
  • FP20 Standard Kit: Suited for wide range of general aqueous samples
  • FP20 Bio Kit: Suited for fast results in aqueous samples such as acids, bases
  • FP20 Micro Kit: Suited for small samples in vials and test tubes
  • FP20 TRIS Kit: Suited for demanding, dirty samples such as emulsions, oily samples, waste water, TRIS
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FiveEasy Plus FP20-TRIS
0.3°C
±0.01
Suitable for wide range of laboratory applications
-2.00 to 16.00
2000 to 2000
-5°C to +105°C
0.1°C
200 measurements, current calibration
22.7 x 14.7 x 7 cm (9 x 5.7 x 2.7 in.)
1.4 lb.
FP20 TRIS Kit
50/60 Hz
FP20 pH meter, LE420 pH sensor, electrode holder, 2 buffer sachets each for pH 4.01, 7, 9.21, and 10.01, test certificate, pH measurement guide
±1
RS232, USB
Linear/Segmented
0.01, 0.1
1mV
23°F to 221°F
LCD
BNC, Cinch, NTC 30kiloOhm sensors
0.63 kg
ATC and MTC
120/240 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers