Learn More
Invitrogen™ FIX & PERM™ Cell Permeabilization Kit
FIX & PERM™ Cell Permeabilization Kit
£92.25 - £254.00
Specifications
Product Type | Fixation/Permeabilization Reagent |
---|---|
Product Line | FIX & PERM™ |
Shipping Condition | Room Temperature |
Product Code | Brand | No. of Tests | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Tests | Price | Quantity & Availability | |||||
10324023
|
Invitrogen™
GAS003 |
50 Tests |
£92.25
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10334023
|
Invitrogen™
GAS004 |
200 Tests |
£254.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The FIX & PERM™ Cell Permeabilization Kit is used in flow cytometry for fixing and permeabilizing cells in suspension, allowing access of antibody to intracellular targets without sacrificing scatter characteristics.The FIX & PERM™ Cell Permeabilization Kit is:
View a selection guide for all available FIX & PERM™ products and products related to sample preparation (including cell lysis and preservation solutions, blocking reagents, and information about the use of Dynabeads™ cell separation products for cell isolation).
The FIX & PERM™ formula was specifically designed to reduce background staining and allow simultaneous addition of fluorochrome-labeled antibodies during the permeabilization step.The FIX & PERM™ Cell Permeabilization Kit is suitable for various biological samples and has been validated for use with cell lines, whole blood, and bone marrow. The FIX & PERM™ kit has been shown to accurately identify previously undetectable intracellular markers. Unlike other fixation and permeabilization reagents available on the market, the FIX & PERM™ staining protocol leaves the morphological scatter characteristics of the cells intact.
How It Works
The FIX & PERM™ Cell Permeabilization Kit contains 2 reagents: Fixation Medium (Medium A) and Permeabilization Medium (Medium B). Cells are incubated for 15 minutes at room temperature with Fixation Medium, washed, and then incubated with the Permeabilization Medium and fluorochrome-labeled antibody. Fixation Medium and Permeabilization Medium are also available separately (Cat. Nos. GAS001S100 & GAS002S100).
Specifications
Fixation/Permeabilization Reagent | |
FIX & PERM™ | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Invitrogen™ FIX & PERM™ Cell Permeabilization Kit