missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fixation Medium (Medium A)
Fixation Medium (Medium A)
£87.00 - £564.00
Specifications
Content And Storage | Store at room temperature. |
---|---|
Product Type | Fixation Medium |
Reagent Type | Fixation Reagent |
Shipping Condition | Room Temperature |
Product Code | Brand | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Price | Quantity & Availability | ||||||
10572625
|
Invitrogen™
GAS001S5 |
5mL |
£87.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10416103
|
Invitrogen™
GAS001S100 |
£564.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fixation Medium (Medium A) is a component of the FIX & PERM™ Cell Permeabilization Kit. The kit is used in flow cytometry for fixing and permeabilizing cells in suspension, allowing access of antibody to intracellular targets without sacrificing scatter characteristics.The FIX & PERM™ Cell Permeabilization Kit is:
View a selection guide for all available FIX & PERM™ products and products related to sample preparation (including cell lysis and preservation solutions, blocking reagents, and information about the use of Dynabeads™ cell separation products for cell isolation).
The Fixation Medium formula was specifically designed to reduce background staining and allow simultaneous addition of fluorochrome-labeled antibodies during the permeabilization step.
How It Works
Cells are incubated for 15 minutes at room temperature with the Fixation Medium and washed. Cells can then be incubated with Permeabilization Medium (Medium B) (Cat. No. GAS002S100) and fluorochrome-labeled antibody. Both Fixation Medium and Permeabilization Medium are included in the FIX & PERM™ Cell Permeabilization Kit.
Specifications
Store at room temperature. | |
Fixation Medium | |
Fixation Reagent | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.