missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Fixed Angle Rotor without Lid for Benchtop Centrifuge

Rotors for Centrifuge 5702/5702 R/5702 RH. The 5702 family of low speed, general purpose centrifuges was especially designed to meet the requirement of low- to mid-throughput clinical research and cell culture labs.

Brand:  Eppendorf™ 5702746002

 View more versions of this product

Product Code. 10663493

  • £419.67 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Rotor F-45-18-17-Cryo

  • Maximum speed: 1,840 × g (4,400 rpm)
  • Quiet centrifugation even without rotor lid
  • Centrifugation of 1 to 2mL Cryo tubes° (max. 13mm dia., use with adapter) and tubes
  • with screw cap (max. 17mm dia., no adapter required)
  • Rotor and adapters are autoclavable (121°C, 20 min.)
  • Rotor F-45-24-11

  • Maximum speed: 1,770 × g (4,400 rpm)
  • Fixed-angle rotor for 24 × 1.5/2.0 mL tubes
  • Quiet centrifugation even without rotor lid
  • Adapters optionally available for: 0.2mL PCR tubes (5425 715.005); 0.4mL centrifuge tubes (5425 717.008); 0.5mL micro test tubes and Microtainer™ tubes (5425 716.001)
  • Rotor and adapters are autoclavable (121°C, 20 min.)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Fixed Angle Rotor
1,770 x g
4,400 rpm
F-45-24-11
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.