Learn More
Fisherbrand™ Fixed Tube Rack
For use with Fisherbrand™ Multi-Platform Shaker and Microplate Mini Shaker
Brand:  Fisherbrand™ 88861208
 View more versions of this product
Product Code. 15703808
  • £92.75 / Each
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Maximum 2 per platform
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fixed Tube Rack
21
262 x 112 x 140 mm
21 × 30mm dia., 50mL tubes, Tubes (5 mL)
Fisherbrand Multi-Platform Shaker and Microplate Mini Shaker
30 mm