Learn More
Fisherbrand™ Fixed Tube Rack
For use with Fisherbrand™ Multi-Platform Shaker and Microplate Mini Shaker
Brand:  Fisherbrand™ 88861206
 View more versions of this product
Product Code. 15783798
  • £89.85 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Maximum 2 per platform
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fixed Tube Rack
40
262 x 112 x 140 mm
40 x 15 mL Tubes
Fisherbrand™ Multi-Platform Shaker and Microplate Mini Shaker
18 mm