missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Flexible Discharge Tube with Recirculation Valve for Dispensette™ S, S Organic, S Trace Analysis
Manufactured from PTFE with a coiled cord approx. 800mm in length.
£119.00 - £123.00
Product Code | Brand | Diameter (Metric) Outer Neck | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer Neck | For Use With (Equipment) | Price | Quantity & Availability | |||||
15448043
|
BRAND™
708132 |
3 mm | For nominal capacity of 2, 5, 10mL |
£119.00
Each |
In Stock
Log in to see stock. |
|||||
15458043
|
BRAND™
708134 |
4.5 mm | For nominal capacity of 25, 50, 100mL |
£123.00
Each |
In Stock
Log in to see stock. |
|||||
Description
- With safety handle
- Not suitable for HF
- Provided individually
Specifications
PTFE | |
1/Pk. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BRAND™ Flexible Discharge Tube with Recirculation Valve for Dispensette™ S, S Organic, S Trace Analysis